Learn More
Restek™ Rtx™-5 Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Similar to DB-5, SPB-5, HP-5, and Ultra II columns
Manufacturer:  Restek™ 10208
 View more versions of this product
Catalog No. 06-708-527
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Drugs, solvent impurities, pesticides, Hydrocarbons, PCB congeners, essential oils, semivolatiles analysis
G27, G36
-60 to 330/350°C
30m
Crossbond™
Rtx™-5
Fused Silica
350°C
0.1μm
0.1μm
Stainless Steel
5% Diphenyl/95% Dimethyl Polysiloxane
Low polar
0.25mm