Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Rtx™ -5 Capillary Columns - 105m Length
General-purpose, fused silica column for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, semivolatiles.
$1528.00 - $1530.00
Specifications
USP Type | G27, G36 |
---|---|
Type | Rtx-5 (G27) Column |
Max. Temperature | 350°C |
Length (Metric) | 105 m |
Material | Stainless Steel |
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Particle Size | Temperature Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Particle Size | Temperature Range | Price | Quantity & Availability | ||
06-708-654
|
Restek™
10244 |
0.25 mm | 0.5 μm | 0.5 μm | -60°C to +320/350°C |
Each for $1,528.00
|
|
||
06-708-706
|
Restek™
10260 |
0.32 mm | 1 μm | 1 μm | -60°C to +310/350°C |
Each for $1,530.00
|
|
||
06-708-763
|
Restek™
10290 |
0.32 mm | 3 μm | 3 μm | -60°C to +260/300°C |
Each for $1,530.00
|
|
||
06-708-728
|
Restek™
10275 |
0.32 mm | 1.5 μm | 1.5 μm | -60°C to +300/320°C |
Each for $1,530.00
|
|
||
06-708-605
|
Restek™
10229 |
0.25 mm | 0.25 μm | 0.25 μm | -60°C to +320/350°C |
Each for $1,528.00
|
|
||
06-708-705
|
Restek™
10259 |
0.25 mm | 1.0 μm | 1 μm | -60°C to +310/340°C | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
Description
All residual catalysts and low molecular weight fragments are removed from the Rtx-5 polymer, providing a tight monomodal distribution and extremely low bleed.
- Low-polarity phase
- Crossbond diphenyl dimethyl polysiloxane
- Equivalent to USP G27 and G36 phases
Note
DB-5MS is a silarylene-based polymer, similar to Rxi-5Sil MS.
Order Info
Custom lengths and film thicknesses available.
Specifications
G27, G36 | |
350°C | |
Stainless Steel | |
Fused Silica |
Rtx-5 (G27) Column | |
105 m | |
Low polar |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title