Restek™ Rtx™ -5 Capillary Columns - 105m Length Available on GSA/VA Contract for Federal Government customers only.

General-purpose, fused silica column for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, semivolatiles.

Manufacturer: Restek™ 10275

 View more versions of this product

Catalog No. 06-708-728

  Check Availability
Add to Cart

    Specifications

    Temperature Range -60° to +300/320°C
    Product Type Rtx-5 (G27) Column
    Packing Material Rtx™-5
    USP Type G27, G36
    Material Stainless Steel
    Technique Crossbond™
    Stationary Phase 5% Diphenyl/95% Dimethyl Polysiloxane
    Polarity Low polar
    Coating Fused Silica
    Diameter (Metric) Inner 0.32mm
    Max. Temperature 350°C
    Particle Size 1.5μm
    Film Thickness 1.5μm
    For Use With Drugs, solvent impurities, pesticides, Hydrocarbons, PCB congeners, essential oils, semivolatiles analysis
    Length (Metric) 105m

    All residual catalysts and low molecular weight fragments are removed from the Rtx-5 polymer, providing a tight monomodal distribution and extremely low bleed.
    • Low-polarity phase
    • Crossbond diphenyl dimethyl polysiloxane
    • Equivalent to USP G27 and G36 phases
    Note
    DB-5MS is a silarylene-based polymer, similar to Rxi-5Sil MS.

    Order Info

    Custom lengths and film thicknesses available.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit