Learn More
Restek™ RTX™-5 Capillary Columns, 30m, Particle Size 1.00µm or Greater Available on GSA/VA Contract for Federal Government customers only.
General-purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners (e.g., Aroclor mixes), essential oils, semivolatiles
Manufacturer:  Restek™ 102796850
 View more versions of this product
Catalog No. 06-708-738
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Drugs, solvent impurities, pesticides, Hydrocarbons, PCB congeners, essential oils, semivolatiles analysis
G27, G36
-60 to 270/290°C
30m
Crossbond
Rtx-5
Fused Silica
Cage
350°C
5μm
5μm
Stainless Steel
5% Diphenyl/95% Dimethyl Polysiloxane
Low polar
0.53mm