Learn More
Restek™ Rtx™-5 Capillary Columns-60m Length Available on GSA/VA Contract for Federal Government customers only.
Similar to DB–5, SPB–5, HP–5, and Ultra2 columns
Manufacturer:  Restek™ 10272
 View more versions of this product
Catalog No. 06-708-722
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Drugs, solvent impurities, pesticides, Hydrocarbons, PCB congeners, essential oils, semivolatiles analysis
G27, G36
-60 to 310/330°C
60m
Crossbond™
Rtx™-5
Fused Silica
350°C
1.5μm
1.5μm
Stainless Steel
5% Diphenyl/95% Dimethyl Polysiloxane
Low polar
0.32mm