Learn More
Restek™ RTX™-50 Capillary Columns: Standard
Similar to DB-17, DB-17ht, HP-17, HP-50, SPB-50, SP-2250; ideal confirmation column for many environmental applications, including EPA Methods 604, 8060, and 608
$313.00 - $1132.00
Specifications
Coating Fused Silica
Packing Material Rtx™-50
For Use With (Application) Pesticides, Herbicides, Rosin Acids, Phthalate Esters, and Sterols
Material Fused Silica
Polarity Midpolar
View More Specs
Includes: Individual test chromatogram
Products 27
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
06-610-730
View Documents
Restek™
40511
0.18 mm 0.4 μm 20 m
Each for $484.00
Add to cart
06-610-728
View Documents
Restek™
40501
0.18 mm 0.20 μm 10 m
Each for $313.00
Add to cart
06-610-729
View Documents
Restek™
40502
0.18 mm 0.20 μm 20 m
Each for $484.00
Add to cart
06-610-712
View Documents
Restek™
10535
0.25 mm 0.5 μm 15 m
Each for $350.00
Add to cart
06-708-945
View Documents
Restek™
10520
0.25 mm 0.25 µm 15 m
Each for $381.00
Add to cart
06-610-715
View Documents
Restek™
10538
0.25 mm 0.5 μm 30 m
Each for $572.00
Add to cart
06-610-719
View Documents
Restek™
10550
0.25 mm 1 μm 15 m
Each for $350.00
Add to cart
06-708-950
View Documents
Restek™
10523
0.25 mm 0.25 µm 30 m
Each for $594.00
Add to cart
06-610-721
View Documents
Restek™
10553
0.25 mm 1 μm 30 m
Each for $572.00
Add to cart
06-610-711
View Documents
Restek™
10524
0.32 mm 0.25 μm 30 m
Each for $615.00
Add to cart
06-610-713
View Documents
Restek™
10536
0.32 mm 0.5 μm 15 m
Each for $377.00
Add to cart
06-708-981
View Documents
Restek™
10552
0.53 mm 1 µm 15 m
Each for $410.00
Add to cart
06-610-716
View Documents
Restek™
10539
0.32 mm 0.5 μm 30 m
Each for $607.00
Add to cart
06-708-971
View Documents
Restek™
10540
0.53 mm 0.5 µm 30 m
Each for $723.00
Add to cart
06-610-717
View Documents
Restek™
10542
0.32 mm 0.5 μm 60 m
Each for $1,075.00
Add to cart
06-610-720
View Documents
Restek™
10551
0.32 mm 1 μm 15 m
Each for $377.00
Add to cart
06-610-722
View Documents
Restek™
10554
0.32 mm 1 μm 30 m
Each for $607.00
Add to cart
06-708-962
View Documents
Restek™
10526
0.25 mm 0.25 µm 60 m
Each for $1,026.00
Add to cart
06-610-723
View Documents
Restek™
10557
0.32 mm 1 μm 60 m
Each for $1,075.00
Add to cart
06-610-710
View Documents
Restek™
10522
0.53 mm 0.25 μm 15 m
Each for $395.00
Add to cart
06-708-990
View Documents
Restek™
10555
0.53 mm 1 µm 30 m
Each for $761.01
Add to cart
06-610-714
View Documents
Restek™
10537
0.53 mm 0.5 μm 15 m
Each for $395.00
Add to cart
06-610-718
View Documents
Restek™
10543
0.53 mm 0.5 μm 60 m
Each for $1,132.00
Add to cart
06-610-726
View Documents
Restek™
10569
0.53 mm 0.83 μm 30 m
Each for $702.00
Add to cart
06-610-724
View Documents
Restek™
10558
0.53 mm 1 μm 60 m
Each for $1,132.00
Add to cart
06-610-725
View Documents
Restek™
10567
0.53 mm 1.5 μm 15 m
Each for $395.00
Add to cart
06-610-727
View Documents
Restek™
10570
0.53 mm 1.5 μm 30 m
Each for $682.00
Add to cart
Description
Description
Columns only.
  • Crossbond™ 50% methyl/50% phenyl polysiloxane
  • Midpolarity
  • Optimized film thickness produces excellent separation of chlorinated pesticides and meets all resolution criteria for CLP pesticides method
  • Crossbond technology provides low bleed, long life, and excellent thermal stability
  • Increased phenyl content offers unique polarity
  • Not adversely affected by column rinsing
Specifications
Specifications
Fused Silica
Pesticides, Herbicides, Rosin Acids, Phthalate Esters, and Sterols
Midpolar
Rtx™-50
Fused Silica
Crossbond Phenyl Methyl Polysiloxane