Learn More
Restek™ RTX™-50 Capillary Columns: Standard Available on GSA/VA Contract for Federal Government customers only.
Similar to DB-17, DB-17ht, HP-17, HP-50, SPB-50, SP-2250; ideal confirmation column for many environmental applications, including EPA Methods 604, 8060, and 608
Manufacturer:  Restek™ 10540
 View more versions of this product
Catalog No. 06-708-971
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Pesticides, herbicides, Rosin Acids, Phthalate Esters, Sterols analysis
0.5μm
0.5μm
Stainless Steel
100% Methyl Polysiloxane
Mid polar
0.53mm
350°C
40 to 270/290°C
30m
GCCrossbond™
Rtx™-50
Fused Silica