Learn More
Restek™ Rtx™-624 Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Application-specific columns for volatile organic pollutants
Manufacturer:  Restek™ 109726850
 View more versions of this product
Catalog No. 06-709-310
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Volatile organic pollutants analysis
G43
-20 to 240°C
60m
Crossbond™
Rtx™-624
Fused Silica
Cage
240deg;C
1.8μm
1.8μm
Stainless Steel
6% Cyanopropylphenyl/94% Dimethyl Polysiloxane
Low to mid polar
0.32mm