Learn More
Restek™ RTX™-65TG Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Application-specific columns, specially tested for triglycerides
Manufacturer:  Restek™ 170086850
 View more versions of this product
Catalog No. 06-710-309
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Triglycerides analysis
G17
40 to 370°C
30m
Crossbond™
Rtx™-65TG
Fused Silica
Cage
350°C
0.1μm
0.1μm
Stainless Steel
65% Diphenyl/35% Dimethyl Polysiloxane
Mid to high polar
0.25mm