Learn More
Restek™ RTX™-BAC2 Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Separate to baseline all blood alcohol compounds in blood, breath or urine
Manufacturer:  Restek™ 180006850
 View more versions of this product
Catalog No. 06-710-334
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Blood Alcohol analysis
2μm
2μm
Stainless Steel
Proprietary
Fused Silica
Cage
350°C
-20 to 240/260°C
30m
Crossbond™
Rtx™-BAC2
0.53mm