Learn More
Restek™ Rtx™-CLPesticides Capillary Columns
Designed to overcome the coelutions and analyte breakdown typically encountered in organochlorine pesticide and herbicide analyses
$522.00 - $1214.00
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Temperature Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Temperature Range | Price | Quantity & Availability | ||
06-709-405
|
Restek™
11139 |
0.32 mm | 0.5 μm | 30 m | -60°C to +320/340°C |
Each for $1,074.00
|
|
||
06-709-401
|
Restek™
11123 |
0.25 mm | 0.25 μm | 30 m | -60°C to +320/340°C |
Each for $988.50
|
|
||
06-610-886
|
Restek™
42102 |
0.18 mm | 0.18 μm | 20 m | 320°C to 340°C |
Each for $702.50
|
|
||
06-709-407
|
Restek™
11140 |
0.53 mm | 0.5 μm | 30 m | -60°C to +300/320°C |
Each for $1,081.00
|
|
||
06-610-883
|
Restek™
11120 |
0.25 mm | 0.25 μm | 15 m | 320°C to 340°C |
Each for $522.00
|
|
||
06-610-884
|
Restek™
11126 |
0.25 mm | 0.25 μm | 60 m | 320°C to 340°C |
Each for $1,214.00
|
|
||
06-610-885
|
Restek™
11136 |
0.32 mm | 0.5 μm | 15 m | 320°C to 340°C |
Each for $551.00
|
|
||
Description
- Application-specific columns exceeding performance criteria for U.S. EPA methods 8081, 608, CLP
- Low bleed—ideal for GC/ECD or GC/MS analyses
- Baseline separations in less than 25 minutes
- Stable to 340°C
Specifications
Rtx™-CLP | |
Fused Silica |