Learn More
Restek™ RTX™-VGC Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Restek™ 19485
 View more versions of this product
Catalog No. 06-710-419
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Volatiles analysis
3μm
3μm
Stainless Steel
Rtx™-VGC
Fused Silica
350°C
-40 to 240/260°C
30m
GC
Rtx™-VGC
0.53mm