Learn More
Restek™ RTX™-VGC Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Restek™ 19420
 View more versions of this product
Catalog No. 06-710-416
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Volatiles analysis
1.8μm
1.8μm
Stainless Steel
Rtx™-VGC
Fused Silica
350°C
-40 to 240/260°C
60m
GC
Rtx™-VGC
0.32mm