Learn More
Restek™ Rtx™-VMS Capillary Columns: Standard
SDP
Provide complete separation of all U.S. EPA Method 8260 compounds in less than 18 minutes
Supplier:  Restek™ 19974
 View more versions of this product
Catalog No. 06-610-914
Add to Cart
Description
Description
  • Offer lower bleed, better selectivity, and overall faster analysis for separating volatile organic compounds, such as those listed in U.S. EPA Method 8260B
  • The Rtx-VMS stationary phase is a highly stable polymer that provides outstanding analysis of volatile compounds, in combination with sensitive ion traps and Agilent 5973 Mass Spectrometers
  • 0.18 and 0.25 mm ID columns allow sample splitting at the injection port, eliminating the added expense and maintenance of a jet separator
  • A 0.45 mm or 0.53 mm ID column can be directly connected to the purge-and-trap transfer line in a system equipped with a jet separator
Specifications
Specifications
Rtx™-V MS
260°C
-40°C to +240/260°C
75 m
0.53 mm
Volatile Organic Pollutants Analysis
3 μm
Fused Silica
Product Suggestions
Product Suggestions