Learn More
Restek™ RTX™-VRX Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
For the proprietary Crossbond™ phase
Manufacturer:  Restek™ 49315
 View more versions of this product
Catalog No. 06-713-255
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Volatile organic pollutants analysis
1μm
1μm
Stainless Steel
Proprietary
Fused Silica
350°C
-40 to 240/260°C
40m
Crossbond™
Rtx™-VRX
0.18mm