Learn More
Restek™ Rtx™-WAX Capillary Columns: With Guard or Cage Available on GSA/VA Contract for Federal Government customers only.
With excellent selectivity compared to other Carbowax™ columns
Manufacturer:  Restek™ 124556850
 View more versions of this product
Catalog No. 06-709-782
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Alkenols, Glycols, Aldehydes analysis
G14, G15, G16, G20, G39
20 to 240/250°C
30m
Crossbond™
Rtx™-Wax
Fused Silica
Cage
350°C
1μm
1μm
Stainless Steel
Carbowax™ Polyethylene Glycol
Intermediate to high polar
0.53mm