Learn More
Restek™ Rxi™-1ms Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Nonpolar phase, Crossbond™ 100% dimethyl polysiloxane
Manufacturer:  Restek™ 13302
 View more versions of this product
Catalog No. 06-715-400
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Arson accelerants, essential oils, Hydrocarbons, pesticides, PCB congeners, Sulfur compounds, Amines, solvent impurities, simulated distillation, Oxygenates, gasoline range organics, refinery gases analysis
0.18μm
0.18μm
Fused Silica
Dimethyl Polysiloxane
Non polar
0.18mm
G2
-60 to 330/350°C
20m
Crossbond™
Rxi™-1 MS
Fused Silica