Learn More
Restek™ Rxi™-5ms Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Low-polarity phase, Crossbond™ 5% diphenyl/95% dimethyl polysiloxane
Manufacturer:  Restek™ 13454
 View more versions of this product
Catalog No. 06-715-451
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Semivolatiles, Phenols, Amines, residual solvents, drugs of abuse, pesticides, PCB congeners, solvent impurities analysis
1μm
1μm
Fused Silica
Diphenyl Dimethyl Polysiloxane
Low polar
0.32mm
G27
-60 to 330/350°C
30m
Crossbond™
Rxi™-5 MS
Fused Silica