Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Septa for 8mm Screw-Thread Vials
Red PTFE
Supplier: Restek™ 21178
Description
Minimize bleed with preconditioned, low-bleed septa.
Handle septa carefully to prevent contamination
Specifications
Red | |
PTFE, Silicone | |
100 |
8 mm | |
1.4 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction