Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Septa for 8mm Screw-Thread Vials

Red PTFE

Manufacturer:  Restek™ 21795

 View more versions of this product

Catalog No. 06-718-678


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.314 x 0.065 in.
Red, White
100
8mm x 0.065 in.
0.8 x 0.161cm
PTFE/Silicone
Vial Cap Septa
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit