Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ SGE Syringes for ThermoFinnigan™/CTC™/LEAP™ Autosamplers

Designed and tested to meet critical autosampler performance needs

Manufacturer:  Restek™ 24926

 View more versions of this product

Catalog No. 06-712-564


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.5μL
0.5μL
Each
Removable
1.96 in.
0.5BNR-C/F-0.47
Gastight™ Autosampler Syringe
26 ga
Cone
ThermoFinnigan™/CTC/LEAP autosamplers
50mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit