Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ SGE Syringes for ThermoFinnigan™/CTC™/LEAP™ Autosamplers

Designed and tested to meet critical autosampler performance needs

Manufacturer:  Restek™ 24930

 View more versions of this product

Catalog No. 06-712-568


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Removable
1.96 in.
10R-C/F-5/0-0.47C
Each
26 ga.
ThermoFinnigan™/CTC/LEAP autosamplers
50mm
Cone
Gastight™ Autosampler Syringe
10μL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit