Learn More
Restek™ Siltek™-Deactivated Guard Columns Available on GSA/VA Contract for Federal Government customers only.
Analyze active samples accurately
Manufacturer:  Restek™ 10027600
 View more versions of this product
Catalog No. 06-717-538
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Chlorinated pesticide analysis
5m
Fused Silica
6/Pk.
380°C
GC
0.32mm