Learn More
Restek™ Siltek™ Splitless Liners for Thermo™ TRACE™ and Focus™ SSL
For Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets.
Manufacturer:  Restek™ 20943213.5
 View more versions of this product
Catalog No. 06-718-214
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Single taper inlet liner
Siltek Splitless w/Deactivated Wool
0.8cm
Thermo Trace™, 8000 series and focus GCs equipped with SSL inlets
5/Pack
0.3cm
10.5cm
Borosilicate Glass