Restek™ Siltek™ Splitless Liners for Thermo™ TRACE™ and Focus™ SSL
For Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets.
Manufacturer: Restek™ 20944213.25
 View more versions of this product
Catalog No. 06-718-219
  Check Availability
Add to Cart
    Specifications
    Product Type Single taper inlet liner
    Packing Material Siltek Splitless w/Deactivated Wool
    Diameter (Metric) Inner 0.3cm
    Diameter (Metric) Outer 0.8cm
    Length (Metric) 10.5cm
    For Use With (Equipment) Thermo Trace™, 8000 series and focus GCs equipped with SSL inlets
    Liner Material Borosilicate Glass
    Quantity 25/Pack
  • Description & Specifications
Related Products
Product Content Feedback