Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Sky™ 3.5mm ID Single Taper/Gooseneck Inlet Liner: Without Wool

For Shimadzu™ 17A, 2010, and 2014 GCs with split/splitless inlets

Manufacturer:  Restek™ 23321.5

 View more versions of this product

Catalog No. 06-712-183

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Specifications

Sky™ Single taper gooseneck inlet liner
0.35cm
9.5cm
Borosilicate Glass
5/Pack
0.5cm
Shimadzu™ 17A, 2010, and 2014 GCs equipped with split/splitless inlets
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit