Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Sky™ 4.0mm ID Drilled Uniliner Inlet Liners: With Wool

Direct injection liners for Agilent™ GCs

Manufacturer:  Restek™ 23307.1

 View more versions of this product

Catalog No. 06-712-168


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Hole near bottom
Each
0.4cm
7.85cm
Borosilicate Glass
Sky™ drilled Uniliner™ inlet liner
Quartz Wool
0.63cm
Agilent GCs equipped with split/splitless inlets
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit