Learn More
Restek™ Sky™ 4.0mm ID Drilled Uniliner Inlet Liners: Without Wool
Direct injection liners for Agilent™ GCs
Manufacturer:  Restek™ 23306.5
 View more versions of this product
Catalog No. 06-712-164
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Hole near bottom
5/Pack
0.63cm
Agilent GCs equipped with split/splitless inlets
Sky™ drilled Uniliner™ inlet linerr with hole near Bottom
0.4cm
7.85cm
Borosilicate Glass