Learn More
Restek™ Sky™ 4.0mm ID Frit Inlet Liner with Top Taper
For Varian™ 1177 GCs with split/splitless ports
Manufacturer:  Restek™ 23330.5
 View more versions of this product
Catalog No. 06-712-220
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Sky™ top taper inlet liner
Glass frit
0.63cm
Bruker/varian GCs equipped with 1177 inlets
5/Pack
0.4cm
7.85cm
Borosilicate Glass