Learn More
Restek™ Sky™ 4.0mm ID Single Taper/Gooseneck Inlet Liners: With Wool
For use with Varian™ 1177 GCs
Manufacturer:  Restek™ 23332.5
 View more versions of this product
Catalog No. 06-712-193
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Sky™ Single taper gooseneck inlet liner
Quartz Wool
0.65cm
Bruker/varian GCs equipped with 1177 inlets
5/Pack
0.4cm
7.85cm
Borosilicate Glass