Learn More
Restek™ Sky™ 5.0mm ID Precision Inlet Liner with wool
For Thermo Scientific™ TRACE™, 8000 Series and Focus™ GCs equipped with SSL inlets
Manufacturer:  Restek™ 23327.5
 View more versions of this product
Catalog No. 06-712-187
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Sky™ Precision™ inlet liner
Quartz Wool
0.8cm
Thermo Trace™, 8000 series and focus GCs equipped with SSL inlets
5/Pack
0.5cm
10.5cm
Borosilicate Glass