Restek™ SPI Liners for Varian, With Buffer

Manufacturer: Restek™ 20851200.5

 View all options for this product

Catalog No. 06-718-096

  Check Availability
Add to Cart


    Packing Material Wool
    Diameter (Metric) Inner 2.4mm
    Diameter (Metric) Outer 4.6mm
    Length (Metric) 54mm
    For Use With (Equipment) Bruker/varian GCs equipped with SPI inlets
    Liner Material Borosilicate Glass
    Quantity 5/Pack
    Deactivation Wool
    Geometry SPI with Buffer
    Includes Buffer
    Product Type SPI liner

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit