Restek™ SPI Liners for Varian, Without Buffer

For Bruker/Varian GCs equipped with SPI inlets.

Manufacturer: Restek™ 20776

 View more versions of this product

Catalog No. 06-717-917

  Check Availability
Add to Cart

    Specifications

    Product Type SPI straight inlet liner
    Diameter (Metric) Inner 0.53mm
    Diameter (Metric) Outer 4.6mm
    Length (Metric) 54mm
    For Use With (Equipment) Bruker/varian GCs equipped with SPI inlets
    Liner Material Borosilicate Glass
    Deactivation Intermediate Polarity (IP)
    Geometry Straight
    Quantity 5/Pack

    • Geometry: Straight
    • Material: Borosilicate Glass
    • OD: 4.6mm
    • Length: 54mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit