Learn More
Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cup Splitter for Glass Liner
For high or low molecular weight compounds
Manufacturer:  Restek™ 20710210.5
 View more versions of this product
Catalog No. 06-717-811
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Base Deactivated
5/Pack
0.4cm
7.85cm
Borosilicate Glass
Cup splitter inlet liner
Base-Deactivated Wool
0.63cm
Agilent GCs equipped with split/splitless inlets