Learn More
Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cup Splitter for Glass Liner
For high or low molecular weight compounds
Manufacturer:  Restek™ 20709
 View more versions of this product
Catalog No. 06-710-924
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
IP cup splitter inlet liner
0.4cm
7.85cm
Borosilicate Glass
Each
0.63cm
Agilent GCs equipped with split/splitless inlets