Learn More
Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cyclosplitter
For use with dirty samples
Manufacturer:  Restek™ 20708
 View more versions of this product
Catalog No. 06-717-803
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
IP Cyclosplitter™ inlet liner
4.0mm
78.5mm
Borosilicate Glass
25/Pk.
6.3mm
Agilent GCs equipped with split/splitless inlets