Learn More
Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cyclosplitter
For use with dirty samples
Manufacturer:  Restek™ 20706
 View more versions of this product
Catalog No. 06-710-916
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
IP Cyclosplitter™ inlet liner
4.0mm
78.5mm
Borosilicate Glass
Ea.
6.3mm
Agilent GCs equipped with split/splitless inlets