Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Split Liners for Shimadzu GCs
SDP

For Shimadzu GCs equipped with split/splitless inlets.

Supplier:  Restek™ 20977

 View more versions of this product

Catalog No. 06-718-301


Add to Cart

Description

Description

  • Material: Borosilicate Glass
  • Deactivation: Intermediate Polarity (IP)
  • OD: 5mm
Specifications

Specifications

Split Straight Inlet Liner
5/Pk.
1.0 mm
Shimadzu 17 A, 2010, and 2014 GCs Equipped with Split/Splitless Inlets
95 mm
Straight
5 mm
Borosilicate Glass
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit