Learn More
Restek™ Split Liners for Shimadzu GCs
For Shimadzu GCs equipped with split/splitless inlets.
Manufacturer:  Restek™ 20861213.5
 View more versions of this product
Catalog No. 06-718-119
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Single Taper
5/Pack
3.5mm
99mm
Borosilicate Glass
Split Single taper inlet liner
Deactivated Wool
5mm
Shimadzu™ 14A GCs equipped with split/splitless inlets