Learn More
Restek™ Stabilwax™ Capillary Columns, 30m Total Length Available on GSA/VA Contract for Federal Government customers only.
Long column lifetime, even when programming repeatedly up to 260°C (500°F)
Manufacturer:  Restek™ 106696850
 View more versions of this product
Catalog No. 06-709-149
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
FAMEs, flavor compounds, essential oils, solvents, Xylene isomers analysis
1.5μm
1.5μm
30m
Crossbond
Stabilwax™
Fused Silica
5 in. cage
G14, G15, G16, G20, G39
40 to 230/240°C
Carbowax Polymer
Stainless Steel
Carbowax Polyethylene Glycol
Polar
0.53mm