Learn More
Restek™ Stabilwax™ Capillary Columns: 50m Length Available on GSA/VA Contract for Federal Government customers only.
Long column lifetimes, even when programming repeatedly up to 260°C (500°F)
Manufacturer:  Restek™ 10642105
 View more versions of this product
Catalog No. 06-709-086
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
FAMEs, flavor compounds, essential oils, solvents, Xylene isomers analysis
0.5μm
0.5μm
60m
Crossbond
Stabilwax
Fused Silica
N/A
G14, G15, G16, G20, G39
40 to 250/260°C
Carbowax Polymer
Stainless Steel
Carbowax Polyethylene Glycol
Polar
0.32mm