Learn More
Restek™ Stabilwax™ Capillary Columns Available on GSA/VA Contract for Federal Government customers only.
Polyethylene glycol phase resists damage from oxygen or moisture
Manufacturer:  Restek™ 10658
 View more versions of this product
Catalog No. 06-709-125
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
FAMEs, flavor compounds, essential oils, solvents, Xylene isomers analysis
1μm
1μm
60m
Crossbond™
Stabilwax™
Fused Silica
G14, G15, G16, G20, G39
40 to 240/260°C
Carbowax™ Polymer
Stainless Steel
Carbowax™ Polyethylene Glycol
Polar
0.53mm