Learn More
Restek™ Stabilwax™ Capillary Columns, Under 30m Available on GSA/VA Contract for Federal Government customers only.
Long column lifetimes, even when programming repeatedly up to 260°C (500°F)
Manufacturer:  Restek™ 10667107
 View more versions of this product
Catalog No. 06-709-141
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
FAMEs, flavor compounds, essential oils, solvents, Xylene isomers analysis
2μm
2μm
15m
Crossbond
Stabilwax
Fused Silica
Not Applicable
G14, G15, G16, G20, G39
40 to 220/230°C
Carbowax Polymer
Stainless Steel
Carbowax Polyethylene Glycol
Polar
0.53mm