Learn More
Restek™ Stabilwax™-DA Capillary Columns, Part 2: 30m Length Available on GSA/VA Contract for Federal Government customers only.
Reduce adsorption and improve responses for many basic compounds
Manufacturer:  Restek™ 110556850
 View more versions of this product
Catalog No. 06-709-390
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Free (underivatized) Acids, some Inorganic Acids analysis
G25, G35
40 to 240/250°C
Acidic
Stainless Steel
Carbowax Polyethylene Glycol
Polar
0.53mm
250°C
1μm
1μm
30m
Crossbond
Stabilwax-DA
Fused Silica
5 in. Cage