Learn More
Restek™ Stabilwax™-DA Capillary Columns, Part 2: 30m Length Available on GSA/VA Contract for Federal Government customers only.
Reduce adsorption and improve responses for many basic compounds
Manufacturer:  Restek™ 110246850
 View more versions of this product
Catalog No. 06-709-349
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Free (underivatized) Acids, some Inorganic Acids analysis
0.25μm
0.25μm
30m
Crossbond
Stabilwax-DA
Fused Silica
5 in. Cage
G25, G35
40 to 250°C
Acidic
Stainless Steel
Carbowax Polyethylene Glycol
Polar
0.32mm