Learn More
Restek™ Stabilwax™-DA Capillary Columnsto be Available on GSA/VA Contract for Federal Government customers only.
Similar in polarity to DB-FFAP, OV-351, NUKOL FFAP, HP FFAP
Manufacturer:  Restek™ 11038
 View more versions of this product
Catalog No. 06-709-367
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Free (underivatized) Acids, some Inorganic Acids analysis
0.5μm
0.5μm
30m
Crossbond™
Stabilwax™-DA
Fused Silica
G25, G35
40 to 250°C
Acidic
Stainless Steel
Carbowax™ Polyethylene Glycol
Polar
0.25mm