Learn More
Restek™ Stabilwax™-DB Capillary Columns, Part 2 Available on GSA/VA Contract for Federal Government customers only.
Reduce adsorption and improve responses for many basic compounds
Manufacturer:  Restek™ 10857105
 View more versions of this product
Catalog No. 06-709-228
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Underivatized Amines, Alkylamines, Diamines, Triamines, Nitrogen containing Heterocyclics analysis
1μm
1μm
50m
Crossbond
Stabilwax-DB
Fused Silica
Not Applicable
250°C
40 to 210/220°C
Amines and other basic compound
Stainless Steel
Base Deactivated Carbowax™ Polyethylene Glycol
Polar
0.32mm