Learn More
Restek™ Straight Inlet Liners for Splitless Injection
Used for samples containing a narrow molecular weight distribution and for those not prone to thermal decomposition
Manufacturer:  Restek™ 22400
 View more versions of this product
Catalog No. 06-711-897
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
IP splitless straight inlet liner
Deactivated Wool
0.65cm
Agilent GCs equipped with split/splitless inlets
Each
0.4cm
7.85cm
Borosilicate Glass