Learn More
Restek™ Straight Inlet Liners for Splitless Injection
Used for samples containing a narrow molecular weight distribution and for those not prone to thermal decomposition
Manufacturer:  Restek™ 20714
 View more versions of this product
Catalog No. 06-717-832
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
IP splitless straight inlet liner
0.2cm
7.85cm
Borosilicate Glass
25/Pack
0.65cm
Agilent GCs equipped with split/splitless inlets