Restek™ Straight Inlet Liners for Splitless Injection

Used for samples containing a narrow molecular weight distribution and for those not prone to thermal decomposition

Manufacturer: Restek™ 20773202.5

 View more versions of this product

Catalog No. 06-717-906

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Diameter (Metric) Inner 4.0mm
    Diameter (Metric) Outer 6.5mm
    Length (Metric) 7.85cm
    For Use With (Equipment) Agilent GCs equipped with split/splitless inlets
    Quantity 5/Pk.
    For Use With (Application) Trace samples greater than 2uL
    Product Type IP splitless straight inlet liner

    • Useful for splitless mode of operation and slow injections, small (<2µL) sample volumes, and high-boiling point solvent systems
    • Compatible with Agilent™/Finnigan™ columns
    • Liners should be packed with wool or fused silica beads to trap nonvolatile residue and prevent column contamination when analyzing dirty samples; also, packing in wool is recommended to aid in vaporization of high molecular weight compounds and to minimize discrimination
    • O.D. x Length: 6.5 x 78.5mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit